Learn More
Aviva Systems Biology Corp MLX Antibody - C-terminal region (ARP39967_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39967_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MLX. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human MLX. Alternate names: MAD7, MXD7, TCFL4, bHLHd13ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57248