Learn More
Aviva Systems Biology Corp MLXIP Antibody - middle region (ARP35891_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35891_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MLXIP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MLXIP. Alternate names: KIAA0867, MIR, MONDOA, bHLHe36ApplicationWB, IHC. Species reactivity: Dog, Human, Mouse, Rabbit
Catalog No. 50-014-55913