Learn More
New England Biolabs, Inc. MlyI – 1000 units
SDP
Catalog No. 50812342
Encompass
A restriction endonuclease that recognizes the sequence GAGTCNNNNN_^.
Catalog No. 50-812-342 Supplier New England Biolabs, Inc. Supplier No. R0610S
A restriction endonuclease that recognizes the sequence GAGTCNNNNN_^.