Learn More
New England Biolabs, Inc. MlyI – 5000 units
SDP
Catalog No. 50812341
Encompass
A restriction endonuclease that recognizes the sequence GAGTCNNNNN_^.
Catalog No. 50-812-341 Supplier New England Biolabs, Inc. Supplier No. R0610L
A restriction endonuclease that recognizes the sequence GAGTCNNNNN_^.