Learn More
Selleck Chemical LLC MM-102
SDP
Catalog No. 5000004194
Encompass_Preferred
MM-102 (HMTase Inhibitor IX) is a high-affinity peptidomimetic inhibitor of the WDR5/MLL1 protein-protein interaction which bind to WDR5 with Ki 1 nM and IC50 2 4nM This product may precipitate when dissolved in saline solution or PBS solution It is recommended to prepare the stock solution in DMSO solution
Catalog No. 50-000-04194 Supplier Selleck Chemical LLC Supplier No. S726520MG
MM-102 (HMTase Inhibitor IX) is a high-affinity peptidomimetic inhibitor of the WDR5/MLL1 protein-protein interaction which bind to WDR5 with Ki 1 nM and IC50 2 4nM This product may precipitate when dissolved in saline solution or PBS solution It is recommended to prepare the stock solution in DMSO solution