Learn More
Apexbio Technology LLC MM-102, 20mg. Cas: 1417329-24-8 MFCD: MFCD28015099. MLL1 inhibitor,high-affinity peptidomimetic
SDP
Supplier:  Apexbio Technology LLC B158220
Encompass_Preferred
MM-102 is an antagonist of MLL1 with IC50 value of 2.4nM [1].Mixed lineage leukemia 1 (MLL1) is a histone H3 lysine 4 (H3K4) methyltransferase. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5268