Learn More
Selleck Chemical LLC MM-102 2mg 1417329-24-8 HMTase Inhibitor IX
SDP
Supplier:  Selleck Chemical LLC S72652MG
Encompass_Preferred
MM-102 (HMTase Inhibitor IX) is a high-affinity peptidomimetic MLL1 inhibitor with IC50 of 0.4 ?M in a cell-free assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-7980