Learn More
Abcam MM-102 (HMTase Inhibitor I x ), 5MG
Catalog No. 502671219
Encompass_Preferred
MW 669.8 Da, Purity >=98%. MM-102 (HMTase Inhibitor IX) is a potent WDR5/MLL interaction inhibitor (IC50 = 2.4 nM). It has been shown to block MLL1 methyltransferase activity, reducing the expression of HoxA9 and Meis-1 genes, which are both critical MLL1 target genes in MLL1 fusion protein-mediated leukemogenesis. WD-repeat protein 5 (WDR5) is a scaffold protein commonly involved in the formation of nucleosome-modifying protein complexes with histones.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1219 Supplier Abcam Supplier No. AB2870225MG
MW 669.8 Da, Purity >=98%. MM-102 (HMTase Inhibitor IX) is a potent WDR5/MLL interaction inhibitor (IC50 = 2.4 nM). It has been shown to block MLL1 methyltransferase activity, reducing the expression of HoxA9 and Meis-1 genes, which are both critical MLL1 target genes in MLL1 fusion protein-mediated leukemogenesis. WD-repeat protein 5 (WDR5) is a scaffold protein commonly involved in the formation of nucleosome-modifying protein complexes with histones.
The product is subject to the following: Abcam Restricted Use Statement