Learn More
TARGETMOL CHEMICALS INC MM-102 TFA 5MG
SDP
Catalog No. 502877870
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. MM-102 TFA (HMTase Inhibitor IX TFA) is a potent WDR5/MLL interaction inhibitor with an IC50 of 2.4 nM. This compound prevents the interaction between mixed lineage leukemia 1 (MLL1) and WD Trp-Asp repeat domain 5 (WDR5), inhibiting MLL1 H3K4 histone methyltransferase (HMT) activity and down-regulating H3K4me3, which facilitates the epigenetic reprogramming of porcine somatic cell nuclear transfer embryos. Purity 99.3%
Catalog No. 50-287-7870 Supplier TARGETMOL CHEMICALS INC Supplier No. T87685MG
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. MM-102 TFA (HMTase Inhibitor IX TFA) is a potent WDR5/MLL interaction inhibitor with an IC50 of 2.4 nM. This compound prevents the interaction between mixed lineage leukemia 1 (MLL1) and WD Trp-Asp repeat domain 5 (WDR5), inhibiting MLL1 H3K4 histone methyltransferase (HMT) activity and down-regulating H3K4me3, which facilitates the epigenetic reprogramming of porcine somatic cell nuclear transfer embryos. Purity 99.3%