Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mm-102 trifluoroacetate | 1883545-52-5 | 99.7% | 1 ML
SDP

Catalog No. 5000230592
Encompass_Preferred

MM-102 TFA is the trifluoroacetate salt of MM-102, a potent inhibitor of the WDR5/MLL protein-protein interaction, supplied as a ready-to-use 10 mM solution in DMSO for research applications.

  • Potent WDR5/MLL interaction inhibitor (IC50 = 2.4 nM).
  • Supplied as a 10 mM solution in DMSO, 1 mL volume.
  • High purity listed at 99.68% (reported as 99.7%).
  • Molecular weight 783.83 g·mol⁻1; formula C37H50F5N7O6.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-30592 Supplier Medchemexpress LLC Supplier No. HY12220A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MM-102 TFA is the trifluoroacetate salt of MM-102, a potent inhibitor of the WDR5/MLL protein-protein interaction, supplied as a ready-to-use 10 mM solution in DMSO for research applications.

  • Potent WDR5/MLL interaction inhibitor (IC50 = 2.4 nM).
  • Supplied as a 10 mM solution in DMSO, 1 mL volume.
  • High purity listed at 99.68% (reported as 99.7%).
  • Molecular weight 783.83 g·mol⁻1; formula C37H50F5N7O6.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.