Learn More
Medchemexpress LLC MM-102 trifluoroacetate | 1883545-52-5 | 99.7% | 2 MG
SDP
Catalog No. 5000232131
Encompass_Preferred
MM-102 TFA is the trifluoroacetate salt of MM-102, a research reagent that inhibits the WDR5/MLL protein-protein interaction. It is supplied as high-purity solid and as DMSO solution formats for biochemical and cellular studies, with reported potency in WDR5 binding assays.
  • Potent inhibitor of the WDR5/MLL protein-protein interaction.
  • Reported IC50 = 2.4 nM in WDR5 binding assay.
  • High reported purity (99.68%).
  • Available in small solid quantities and as 10 mM in DMSO solution.
  • Trifluoroacetate salt form for convenient handling and solubility.
Catalog No. 50-002-32131 Supplier Medchemexpress LLC Supplier No. HY12220A2MG
MM-102 TFA is the trifluoroacetate salt of MM-102, a research reagent that inhibits the WDR5/MLL protein-protein interaction. It is supplied as high-purity solid and as DMSO solution formats for biochemical and cellular studies, with reported potency in WDR5 binding assays.
  • Potent inhibitor of the WDR5/MLL protein-protein interaction.
  • Reported IC50 = 2.4 nM in WDR5 binding assay.
  • High reported purity (99.68%).
  • Available in small solid quantities and as 10 mM in DMSO solution.
  • Trifluoroacetate salt form for convenient handling and solubility.