Learn More
Abcam MM 11253, RARgamma, 10MG
Catalog No. 502706549
Encompass_Preferred
MW 462.66 g/mol, Purity >98%. Selective RARγ antagonist. Blocks the growth inhibitory effects of RARγ-selective agonists. Shows antiapoptotic effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6549 Supplier Abcam Supplier No. AB14150810MG
MW 462.66 g/mol, Purity >98%. Selective RARγ antagonist. Blocks the growth inhibitory effects of RARγ-selective agonists. Shows antiapoptotic effects.
The product is subject to the following: Abcam Restricted Use Statement