Learn More
Enzo Life Sciences MM 4-64 (1mg)
SDP
Supplier:  Enzo Life Sciences ENZ52252
Encompass_Preferred
MM 4-64 is a cationic dye, equivalent to membrane probe FM 4-64. MM 4-64 is a lipophilic styryl dye that is used as a vital stain to follow bulk membrane-internalization and transport to the vacuole in yeast. MM 4-64 is also a sensitive reporter of vacuolar dynamics, detecting such events as segregation structure formation during mitosis, vacuole fission/fusion events, and vacuolar morphology in different classes of vacuolar protein sorting mutants. Wavelength Maxima: Excitation 558nm, Emission 734nm Alternative name: N-(3-Triethylammoniumpropyl)-4-(6-(4-(diethylamino)phenyl)hexatrienyl)pyridinium dibromide. Purity: ≥90% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-2628