Learn More
AdipoGen MM 47755
SDP
Supplier:  AdipoGen BVT0282M001
Encompass_Preferred
Chemical. CAS 117620-87-8. Formula C20H16O5. MW 336.3. Isolated from Streptomyces sp. Gö 40/14. Antibiotic. Angucyclinone. Antibacterial and antifungal.
Catalog No. 50-256-2020