Learn More
Medchemexpress LLC Mm-589 (racemic mixture) TFA | 2097887-20-0 | 686.72 | C30H45F3N8O7 | 10 MG
SDP
Catalog No. 5000211723
Encompass_Preferred
MM-589 (racemic mixture) TFA is a small-molecule inhibitor of the WDR5-MLL protein-protein interaction supplied as a racemic trifluoroacetic acid salt for research use. It is intended for biochemical and cellular studies targeting WDR5-mediated epigenetic regulation.
  • Small-molecule inhibitor of WDR5-MLL protein-protein interaction.
  • Racemic mixture supplied as a trifluoroacetic acid (TFA) salt.
  • Intended for biochemical and cellular research applications.
  • Molecular formula C30H45F3N8O7 and molecular weight 686.72 (TFA salt).
  • Available in a 10 mg research pack size.
Catalog No. 50-002-11723 Supplier Medchemexpress LLC Supplier No. HY100869B10MG
Add to Cart
Add to Cart
MM-589 (racemic mixture) TFA is a small-molecule inhibitor of the WDR5-MLL protein-protein interaction supplied as a racemic trifluoroacetic acid salt for research use. It is intended for biochemical and cellular studies targeting WDR5-mediated epigenetic regulation.
  • Small-molecule inhibitor of WDR5-MLL protein-protein interaction.
  • Racemic mixture supplied as a trifluoroacetic acid (TFA) salt.
  • Intended for biochemical and cellular research applications.
  • Molecular formula C30H45F3N8O7 and molecular weight 686.72 (TFA salt).
  • Available in a 10 mg research pack size.