Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mm-589 (racemic mixture) TFA | 99.8% | 686.72 g/mol | C30H45F3N8O7 | 25 MG
SDP

Catalog No. 5000211479
Encompass_Preferred

MM-589 (racemic mixture) TFA is the trifluoroacetic acid salt of a macrocyclic peptidomimetic inhibitor that targets the WDR5-MLL protein-protein interaction. Supplied as a high-purity solid for research use, it is intended for biochemical and cell-based assays; handle sealed and protected from moisture, and follow recommended cold storage conditions for stability.

  • Potent inhibitor of WDR5-MLL protein-protein interaction.
  • Supplied as a racemic TFA salt in solid powder form.
  • High purity, 99.83%.
  • Molecular weight 686.72 g/mol; formula C30H45F3N8O7.
  • Available in small research package sizes, including 25 mg.
  • Recommended storage: in solvent -80°C for up to 6 months, or -20°C for up to 1 month (sealed).

Catalog No. 50-002-11479 Supplier Medchemexpress LLC Supplier No. HY100869B25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MM-589 (racemic mixture) TFA is the trifluoroacetic acid salt of a macrocyclic peptidomimetic inhibitor that targets the WDR5-MLL protein-protein interaction. Supplied as a high-purity solid for research use, it is intended for biochemical and cell-based assays; handle sealed and protected from moisture, and follow recommended cold storage conditions for stability.

  • Potent inhibitor of WDR5-MLL protein-protein interaction.
  • Supplied as a racemic TFA salt in solid powder form.
  • High purity, 99.83%.
  • Molecular weight 686.72 g/mol; formula C30H45F3N8O7.
  • Available in small research package sizes, including 25 mg.
  • Recommended storage: in solvent -80°C for up to 6 months, or -20°C for up to 1 month (sealed).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.