Learn More
World Precision Instrument MM-6 Micropositioner-505083
SDP
Catalog No. 502539259
Encompass
The MM-6 micropositioner is light weight, accurate, compact device designed to eliminate many research and design problems, especially in the case of space limitations. Hardened steel precision pre-loaded ball bearing slides and high resolution thumbscrew movement assure smooth motion, precision, and excellent load bearing capacity.
The black anodized aluminum body is made with a one-piece base and three piece construction. The top plate is designed around an M2 thread size on a 10 mm square hole pattern and central M4 tapped hole. The MM-6 is available in single, double, or triple stages. Adding stages is an easy job and requires no special tools. Triple stages come with an angle bracket to attach the Z-axis. The travel of the stage is 0.26" (6.5 mm) and the base is easily adapted to most surfaces.
Catalog No. 50-253-9259 Supplier World Precision Instrument Supplier No. 505083
The MM-6 micropositioner is light weight, accurate, compact device designed to eliminate many research and design problems, especially in the case of space limitations. Hardened steel precision pre-loaded ball bearing slides and high resolution thumbscrew movement assure smooth motion, precision, and excellent load bearing capacity.
The black anodized aluminum body is made with a one-piece base and three piece construction. The top plate is designed around an M2 thread size on a 10 mm square hole pattern and central M4 tapped hole. The MM-6 is available in single, double, or triple stages. Adding stages is an easy job and requires no special tools. Triple stages come with an angle bracket to attach the Z-axis. The travel of the stage is 0.26" (6.5 mm) and the base is easily adapted to most surfaces.