Learn More
Apexbio Technology LLC Mouse Tissue Lysis Kit 200rxns
SDP
Catalog No. 503107438
Encompass_Preferred
The Mouse Tissue Lysis Kit is designed for the first step (tissue digestion) in experiments using the Direct Mouse Genotyping Kit The lysis buffer and neutralization buffer rapidly digest mouse tissues (tail toe or ear) to efficiently release intact genomic DNA DNA extracted by this method can be used directly as a PCR template without further purification steps significantly reducing digestion time
Catalog No. 50-310-7438 Supplier Apexbio Technology LLC Supplier No. K1038200
The Mouse Tissue Lysis Kit is designed for the first step (tissue digestion) in experiments using the Direct Mouse Genotyping Kit The lysis buffer and neutralization buffer rapidly digest mouse tissues (tail toe or ear) to efficiently release intact genomic DNA DNA extracted by this method can be used directly as a PCR template without further purification steps significantly reducing digestion time