Learn More
ATCC MM.1R; Myeloma; Human (Homo sapiens)
Catalog No. 502382485
Encompass
MM.1R is a B lymphoblast cell line that was isolated in 1990 from the peripheral blood of a 42-year-old, Black female with multiple myeloma who had become resistant to steroid-based therapy. It has applications for immune system disorder research and immunology.
Catalog No. 50-238-2485 Supplier ATCC Supplier No. CRL2975
MM.1R is a B lymphoblast cell line that was isolated in 1990 from the peripheral blood of a 42-year-old, Black female with multiple myeloma who had become resistant to steroid-based therapy. It has applications for immune system disorder research and immunology.