Learn More
Aviva Systems Biology Corp MMAA Antibody - N-terminal region (ARP64020_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64020_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MMAA. It was validated on Western Blot. . Alternate names: MGC120010, MGC120011, MGC120012, MGC120013, cblAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-288-144