Learn More
Medchemexpress LLC HY-15581 5mg Medchemexpress, MMAD CAS:203849-91-6 Purity:>98%
SDP
Catalog No. 502081743
Encompass_Preferred
Medchemexpress, HY-15581 5mg MMAD CAS:203849-91-6 MMAD is a potent tubulin inhibitor, is a toxin payload in antibody drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1743 Supplier Medchemexpress LLC Supplier No. HY155815MG
Medchemexpress, HY-15581 5mg MMAD CAS:203849-91-6 MMAD is a potent tubulin inhibitor, is a toxin payload in antibody drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.