Learn More
Medchemexpress LLC MMAD-d8 | 203849-91-6 | 99.0% | 779.11 | C41H58D8N6O6S | 10 MG
SDP
Catalog No. 5000286830
Encompass_Preferred
MMAD-d8 is the deuterated form of monomethyl dolastatin 10 (MMAD), a potent microtubule-disrupting agent provided for research use. It is used primarily as a cytotoxic payload in antibody-drug conjugate (ADC) development and for tracer or stability studies where deuteration improves analytical utility. The compound is supplied as a solid powder and is unstable in solution, so solutions should be prepared immediately before use.
Catalog No. 50-002-86830 Supplier Medchemexpress LLC Supplier No. HY15581S10MG
MMAD-d8 is the deuterated form of monomethyl dolastatin 10 (MMAD), a potent microtubule-disrupting agent provided for research use. It is used primarily as a cytotoxic payload in antibody-drug conjugate (ADC) development and for tracer or stability studies where deuteration improves analytical utility. The compound is supplied as a solid powder and is unstable in solution, so solutions should be prepared immediately before use.