Learn More
Medchemexpress LLC MMAE-SMCC | 2021179-11-1 | 95.7% | 1140.43 | C58H89N7O14S | 5 MG
SDP
Catalog No. 5000202964
Encompass_Preferred
MMAE-SMCC is an antibody-drug conjugate (ADC) linker-payload conjugate that couples the cytotoxic agent monomethyl auristatin E (MMAE) to an SMCC linker for ADC research applications. It is supplied as a dry powder for research use only, with reported purity of approximately 95.65%, molecular formula C58H89N7O14S, and molecular weight 1140.43. Store at -20 °C, protect from light, and store under nitrogen. The compound is unstable in solution; prepare fresh solutions before use.
  • Composed of monomethyl auristatin E linked via an SMCC linker.
  • High purity suitable for research (95.7%).
  • Supplied as a dry powder; store at -20 °C and protect from light.
  • Unstable in solution; prepare fresh solutions prior to use.
  • Intended for research use only.
Catalog No. 50-002-02964 Supplier Medchemexpress LLC Supplier No. HY1356605MG
MMAE-SMCC is an antibody-drug conjugate (ADC) linker-payload conjugate that couples the cytotoxic agent monomethyl auristatin E (MMAE) to an SMCC linker for ADC research applications. It is supplied as a dry powder for research use only, with reported purity of approximately 95.65%, molecular formula C58H89N7O14S, and molecular weight 1140.43. Store at -20 °C, protect from light, and store under nitrogen. The compound is unstable in solution; prepare fresh solutions before use.
  • Composed of monomethyl auristatin E linked via an SMCC linker.
  • High purity suitable for research (95.7%).
  • Supplied as a dry powder; store at -20 °C and protect from light.
  • Unstable in solution; prepare fresh solutions prior to use.
  • Intended for research use only.