Learn More
Medchemexpress LLC HY-15579A 10mg , MMAF (Hydrochloride) Monomethylauristatin F Hydrochloride CAS:1415246-68-2 Purity:98%
SDP
Catalog No. 501494078
Encompass_Preferred
Medchemexpress, HY-15579A 10mg MMAF (Hydrochloride) Monomethylauristatin F Hydrochloride CAS:1415246-68-2 Formula:C39H66ClN5O8 IC50: 119 nM (Cytotoxicity, Karpas 299 cell), 105 nM (Cytotoxicity, H3396 cell), 257 nM (Cytotoxicity, 786-O cell), 200 nM (Cytotoxicity, Caki-1, cell) Purity:98% MMAF hydrochloride is an antitubulin agent that inhibit cell division; inhibits H3397 cell growth with an IC 50 of 105 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-4078 Supplier Medchemexpress LLC Supplier No. HY15579A10MG
Medchemexpress, HY-15579A 10mg MMAF (Hydrochloride) Monomethylauristatin F Hydrochloride CAS:1415246-68-2 Formula:C39H66ClN5O8 IC50: 119 nM (Cytotoxicity, Karpas 299 cell), 105 nM (Cytotoxicity, H3396 cell), 257 nM (Cytotoxicity, 786-O cell), 200 nM (Cytotoxicity, Caki-1, cell) Purity:98% MMAF hydrochloride is an antitubulin agent that inhibit cell division; inhibits H3397 cell growth with an IC 50 of 105 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.