Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MMAF-OtBu (tert-butyl ester of monomethylauristatin F) | 745017-95-2 | 99.8% | 788.07 | C43H73N5O8 | 25 MG
SDP

Catalog No. 5000204455
Encompass_Preferred

MMAF-OtBu is a tert-butyl-protected derivative of monomethylauristatin F used as a cytotoxic payload building block for the synthesis of antibody-drug conjugates (ADCs). Supplied as a white to off-white solid, the compound is supplied in small milligram pack sizes and has defined storage conditions to maintain stability.

  • Tert-butyl-protected monomethylauristatin F derivative.
  • Used as a cytotoxic building block in ADC synthesis.
  • Chemical formula C43H73N5O8; molecular weight 788.07.
  • Purity 99.8%.
  • Solid, white to off-white; store at -20°C; in solvent: -80°C (6 months), -20°C (1 month).
  • Available in small milligram pack sizes for research and conjugation workflows.

Catalog No. 50-002-04455 Supplier Medchemexpress LLC Supplier No. HY15622525MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MMAF-OtBu is a tert-butyl-protected derivative of monomethylauristatin F used as a cytotoxic payload building block for the synthesis of antibody-drug conjugates (ADCs). Supplied as a white to off-white solid, the compound is supplied in small milligram pack sizes and has defined storage conditions to maintain stability.

  • Tert-butyl-protected monomethylauristatin F derivative.
  • Used as a cytotoxic building block in ADC synthesis.
  • Chemical formula C43H73N5O8; molecular weight 788.07.
  • Purity 99.8%.
  • Solid, white to off-white; store at -20°C; in solvent: -80°C (6 months), -20°C (1 month).
  • Available in small milligram pack sizes for research and conjugation workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.