Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MMAF sodium | 1799706-65-2 | 99.6% | 753.94 g/mol | C39H64N5NaO8 | 50 MG
SDP

Catalog No. 5000204424
Encompass_Preferred

MMAF sodium is a potent tubulin polymerization inhibitor used as a cytotoxic payload in antibody-drug conjugate research. It is supplied as a high-purity, white to off-white solid with documented solubility in common solvents; solutions should be prepared fresh due to instability.

  • Potent tubulin polymerization inhibitor for ADC research.
  • High purity (99.6%) and white to off-white solid appearance.
  • Molecular weight 753.94 g/mol; formula C39H64N5NaO8.
  • Soluble in DMSO (≥ 200 mg/mL) and water (≥ 100 mg/mL).
  • Store at 4°C, protected from light; prepare fresh solutions due to solution instability.

Catalog No. 50-002-04424 Supplier Medchemexpress LLC Supplier No. HY15579B50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MMAF sodium is a potent tubulin polymerization inhibitor used as a cytotoxic payload in antibody-drug conjugate research. It is supplied as a high-purity, white to off-white solid with documented solubility in common solvents; solutions should be prepared fresh due to instability.

  • Potent tubulin polymerization inhibitor for ADC research.
  • High purity (99.6%) and white to off-white solid appearance.
  • Molecular weight 753.94 g/mol; formula C39H64N5NaO8.
  • Soluble in DMSO (≥ 200 mg/mL) and water (≥ 100 mg/mL).
  • Store at 4°C, protected from light; prepare fresh solutions due to solution instability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.