Learn More
Avanti Polar Lipids MMBRNE SUPPORT FOR MINI EXTRDR
Catalog No. NC0419716
Encompass
PTFE Internal Membrane Supports with O-rings (2)
Catalog No. NC0419716 Supplier Avanti Polar Lipids Supplier No. 6100251EA
PTFE Internal Membrane Supports with O-rings (2)