Learn More
Medchemexpress LLC Mmc (TMZ)-TOC | >98.0% | 1725.82 | C74H99F3N20O21S2 | 10 MG
SDP
Catalog No. 5000289084
Encompass_Preferred
MMC(TMZ)-TOC TFA is a temozolomide delivery conjugate designed to selectively bind somatostatin receptor subtype-2 (SSTR2) and deliver TMZ to SSTR2-positive tumor cells for targeted cancer research applications.
  • Targets somatostatin receptor subtype-2 (SSTR2) for receptor-mediated delivery.
  • Conjugated with temozolomide to enable targeted chemotherapy delivery.
  • Suitable for in vitro and in vivo cancer research studies.
  • Provided as a trifluoroacetic acid (TFA) salt for improved handling and stability.
  • Reported purity greater than 98%, suitable for research use.
  • Available in small research quantities for preclinical experiments.
Catalog No. 50-002-89084 Supplier Medchemexpress LLC Supplier No. HY163317A10MG
MMC(TMZ)-TOC TFA is a temozolomide delivery conjugate designed to selectively bind somatostatin receptor subtype-2 (SSTR2) and deliver TMZ to SSTR2-positive tumor cells for targeted cancer research applications.
  • Targets somatostatin receptor subtype-2 (SSTR2) for receptor-mediated delivery.
  • Conjugated with temozolomide to enable targeted chemotherapy delivery.
  • Suitable for in vitro and in vivo cancer research studies.
  • Provided as a trifluoroacetic acid (TFA) salt for improved handling and stability.
  • Reported purity greater than 98%, suitable for research use.
  • Available in small research quantities for preclinical experiments.