Learn More
Medchemexpress LLC MME Human HEK293 100ug
SDP
Catalog No. 5000181865
Encompass_Preferred
MME proteins exhibit thermolysin-like specificity and primarily degrade polypeptides of up to 30 amino acids Crucially it plays a key role in cleaving the Gly-Phe bond to degrade opioid peptides including Met-enkephalin and Leu-enkephalin MME Protein Human (HEK293) is the recombinant human-derived MME protein expressed by HEK293 with tag free
Catalog No. 50-001-81865 Supplier Medchemexpress LLC Supplier No. HYP72876A100UG
MME proteins exhibit thermolysin-like specificity and primarily degrade polypeptides of up to 30 amino acids Crucially it plays a key role in cleaving the Gly-Phe bond to degrade opioid peptides including Met-enkephalin and Leu-enkephalin MME Protein Human (HEK293) is the recombinant human-derived MME protein expressed by HEK293 with tag free