Learn More
Medchemexpress LLC MME Human HEK293 100ug
Supplier: Medchemexpress LLC HYP72876100UG
MME proteins exhibit thermolysin-like specificity and primarily degrade polypeptides of up to 30 amino acids Crucially it plays a key role in cleaving the Gly-Phe bond to degrade opioid peptides including Met-enkephalin and Leu-enkephalin MME Protein Human (HEK293) is the recombinant human-derived MME protein expressed by HEK293 with tag free