Learn More
New England Biolabs, Inc. MmeI – 100 units
SDP
Catalog No. 50812388
Encompass
A restriction endonuclease that recognizes the sequence TCCRACNNNNNNNNNNNNNNNNNN_NN^.
Catalog No. 50-812-388 Supplier New England Biolabs, Inc. Supplier No. R0637S
A restriction endonuclease that recognizes the sequence TCCRACNNNNNNNNNNNNNNNNNN_NN^.