Learn More
New England Biolabs, Inc. MmeI – 500 units
SDP
Catalog No. 50812387
Encompass
A restriction endonuclease that recognizes the sequence TCCRACNNNNNNNNNNNNNNNNNN_NN^.
Catalog No. 50-812-387 Supplier New England Biolabs, Inc. Supplier No. R0637L
Add to Cart
Add to Cart
A restriction endonuclease that recognizes the sequence TCCRACNNNNNNNNNNNNNNNNNN_NN^.