Learn More
Aviva Systems Biology Corp MMEL1 Antibody - middle region (ARP50190_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50190_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MMEL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MMEL1. Alternate names: MGC119454, MGC119455, MGC119456, MMEL2, NEPII, NL1, NL2, SEP, NEP2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-60280