Learn More
AdipoGen MMG-0358 (5 mg)
SDP
Catalog No. 502054002
Encompass_Preferred
Chemical. CAS: 1378976-02-3. Formula: C8H6ClN3O. MW: 195.6. Potent triazole IDO1 (indoleamine 2,3-dioxygenase 1) inhibitor. Has IC50 values of 2nM in a cellular assay on mIDO1, 80nM in a cellular assay on hIDO1, 330nM in an enzymatic assay on hIDO1 at pH 6.5 and 71nM in an enzymatic assay on hIDO1 at pH 7.4. Shown to be active in vivo.
Catalog No. 50-205-4002 Supplier AdipoGen Supplier No. AGCR13630M005
Add to Cart
Add to Cart
Chemical. CAS: 1378976-02-3. Formula: C8H6ClN3O. MW: 195.6. Potent triazole IDO1 (indoleamine 2,3-dioxygenase 1) inhibitor. Has IC50 values of 2nM in a cellular assay on mIDO1, 80nM in a cellular assay on hIDO1, 330nM in an enzymatic assay on hIDO1 at pH 6.5 and 71nM in an enzymatic assay on hIDO1 at pH 7.4. Shown to be active in vivo.