Learn More
LGC GENOMICS LLC (Lucigen) MMLV High Performance Reverse Transcriptase, 25,000 Units @200 U/LOptimised reverse transcriptase and buffer system for the production of full-length cDNA.
Catalog No. NC1896599
Encompass
The MMLV High Performance Reverse Transcriptase produces full-length first-strand cDNA from total RNA preparations or purified poly(A+) RNA.
Catalog No. NC1896599 Supplier LGC GENOMICS LLC Supplier No. RT80125K
The MMLV High Performance Reverse Transcriptase produces full-length first-strand cDNA from total RNA preparations or purified poly(A+) RNA.