Learn More
LGC GENOMICS LLC (Lucigen) MMLV Reverse Transcriptase 1St-Strand cDNA Synthesis Kit, 50 RxnsOptimised, complete system for production of full-length cDNA from low amounts of total RNA.
Supplier:  LGC GENOMICS LLC MM070150
Encompass
Synthesize full-length cDNA (15 kb)Generate first-strand cDNA from picogram amounts of total RNASelect oligo(dT) or random primers - both includedKit contains MMLV Reverse Transcriptase, 10X Reaction Buffer, RiboGuard RNase Inhibitor, dNTP PreMix, DTT, Oligo(dT)21 Primer, Random 9-mer Primers, RNase-free Water.
Catalog No. NC2162065