Learn More
Meridian Life Science MMLV-RT, 200UL
SDP
Catalog No. 5000856230
Encompass_Preferred
MMLV-RT is a high-quality reverse transcriptase that synthesizes a complementary DNA (cDNA) strand from mRNA or total RNA. With higher thermostability and reduced RNase H activity, it can be used for synthesis with long messenger RNA templates with high sensitivity and efficiency and is ideal for detecting low-level target genes.
Catalog No. 50-008-56230 Supplier Meridian Life Science Supplier No. MDX04402
MMLV-RT is a high-quality reverse transcriptase that synthesizes a complementary DNA (cDNA) strand from mRNA or total RNA. With higher thermostability and reduced RNase H activity, it can be used for synthesis with long messenger RNA templates with high sensitivity and efficiency and is ideal for detecting low-level target genes.