Learn More
Meridian Life Science MMLV-RT, 20ML
SDP
Catalog No. 5000856425
Encompass
MMLV-RT is a high-quality reverse transcriptase that synthesizes a complementary DNA (cDNA) strand from mRNA or total RNA. With higher thermostability and reduced RNase H activity, it can be used for synthesis with long messenger RNA templates with high sensitivity and efficiency and is ideal for detecting low-level target genes.
Catalog No. 50-008-56425 Supplier Meridian Life Science Supplier No. MDX04420
MMLV-RT is a high-quality reverse transcriptase that synthesizes a complementary DNA (cDNA) strand from mRNA or total RNA. With higher thermostability and reduced RNase H activity, it can be used for synthesis with long messenger RNA templates with high sensitivity and efficiency and is ideal for detecting low-level target genes.