Learn More
Enzo Life Sciences MMP-1 Assay Buffer (20ml)
SDP
Catalog No. 502011104
Encompass_Preferred
Formulation: Liquid. In 50mM HEPES, pH 7.5, 10mM CaCl2 and 0.05% Brij-35. Long Term Storage: Ambient.
Catalog No. 50-201-1104 Supplier Enzo Life Sciences Supplier No. BMLKI1750020
Add to Cart
Add to Cart
Formulation: Liquid. In 50mM HEPES, pH 7.5, 10mM CaCl2 and 0.05% Brij-35. Long Term Storage: Ambient.