Learn More
CPC Scientific DNP-Pro-Cha-Gly-Cys(Me)-His-Ala-Lys(Nma)-NH2 (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502109747
Encompass
SEQUENCE: DNP-Pro-Cha-Gly-Cys(Me)-His-Ala-Lys(Nma)-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Dnp-P-Cha-G-Cys(Me)-HA-K(Nma)-NH2MOLECULAR FORMULA: C49H68N14O12SMOLECULAR WEIGHT: 1077.22STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [150956-92-6]SYNONYMS: Dnp-Pro--cyclohexyl-Ala-Gly-Cys(Me)-His-Ala-Lys(N-Me-Abz)-NH2RESEARCH AREA: CancerCATEGORY: MMP Substrates REFERENCES:J. Berman et al., J. Biol. Chem., 267, 1434 (1992)SKU(s): MMPS-002A, MMPS-002B, MMPS-002C, MMPS-002D
Catalog No. 50-210-9747 Supplier CPC Scientific Supplier No. MMPS002A
SEQUENCE: DNP-Pro-Cha-Gly-Cys(Me)-His-Ala-Lys(Nma)-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Dnp-P-Cha-G-Cys(Me)-HA-K(Nma)-NH2MOLECULAR FORMULA: C49H68N14O12SMOLECULAR WEIGHT: 1077.22STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [150956-92-6]SYNONYMS: Dnp-Pro--cyclohexyl-Ala-Gly-Cys(Me)-His-Ala-Lys(N-Me-Abz)-NH2RESEARCH AREA: CancerCATEGORY: MMP Substrates REFERENCES:J. Berman et al., J. Biol. Chem., 267, 1434 (1992)SKU(s): MMPS-002A, MMPS-002B, MMPS-002C, MMPS-002D