Learn More
CPC Scientific DNP-Pro-Leu-Ala-Leu-Trp-Ala-Arg-OH (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502109751
Encompass
ONE-LETTER SEQUENCE: DNP-PLALWARMOLECULAR FORMULA: C46H65N13O12MOLECULAR WEIGHT:992.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: CancerREFERENCES: S. Netzel-Arnett, et al., Anal. Biochem. 195, 86 (1991)
Catalog No. 50-210-9751 Supplier CPC Scientific Supplier No. MMPS003A
ONE-LETTER SEQUENCE: DNP-PLALWARMOLECULAR FORMULA: C46H65N13O12MOLECULAR WEIGHT:992.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: CancerREFERENCES: S. Netzel-Arnett, et al., Anal. Biochem. 195, 86 (1991)