Learn More
CPC Scientific DNP-Pro-Leu-Gly-Cys(Me)-His-Ala-DArg-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502109754
Encompass
Efficient substrate for interstitial/vertebrate collagenase (MMP-1), stromelysin (MMP-3), gelatinases (MMP-2 and MMP-9) and punctuated metalloproteinase (MMP-7). ONE-LETTER SEQUENCE: Dnp-PLG-Cys(Me)-HA-DArg-NH2MOLECULAR FORMULA: C38H57N15O11S1MOLECULAR WEIGHT:932.03STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [145224-98-2], SYNONYMS: Dnp-Pro-Leu-Gly-Cys(Me)-His-Ala-D-Arg-NHRESEARCH AREA: CancerREFERENCES: J. Berman et al, JBC, 267, 1434 (1992)
Catalog No. 50-210-9754 Supplier CPC Scientific Supplier No. MMPS004B
Efficient substrate for interstitial/vertebrate collagenase (MMP-1), stromelysin (MMP-3), gelatinases (MMP-2 and MMP-9) and punctuated metalloproteinase (MMP-7). ONE-LETTER SEQUENCE: Dnp-PLG-Cys(Me)-HA-DArg-NH2MOLECULAR FORMULA: C38H57N15O11S1MOLECULAR WEIGHT:932.03STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [145224-98-2], SYNONYMS: Dnp-Pro-Leu-Gly-Cys(Me)-His-Ala-D-Arg-NHRESEARCH AREA: CancerREFERENCES: J. Berman et al, JBC, 267, 1434 (1992)