Learn More
CPC Scientific DNP-Pro-Cha-Abu-Cys(Me)-His-Ala-Lys(Nma)-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502109756
Encompass
SEQUENCE: DNP-Pro-Cha-Abu-Cys(Me)-His-Ala-Lys(Nma)-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Dnp-P-Cha-Abu-Cys(Me)-HA-K(Nma)-NH2MOLECULAR FORMULA: C51H12N14O12S1MOLECULAR WEIGHT: 1105.29STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [150956-93-7]SYNONYMS: Dnp-Pro-Cha-Abu-Cys(Me)-His-Ala-Lys(N-Me-Abz)-NHRESEARCH AREA: CancerREFERENCES:SKU(s): MMPS-005A, MMPS-005B
Catalog No. 50-210-9756 Supplier CPC Scientific Supplier No. MMPS005B
SEQUENCE: DNP-Pro-Cha-Abu-Cys(Me)-His-Ala-Lys(Nma)-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Dnp-P-Cha-Abu-Cys(Me)-HA-K(Nma)-NH2MOLECULAR FORMULA: C51H12N14O12S1MOLECULAR WEIGHT: 1105.29STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [150956-93-7]SYNONYMS: Dnp-Pro-Cha-Abu-Cys(Me)-His-Ala-Lys(N-Me-Abz)-NHRESEARCH AREA: CancerREFERENCES:SKU(s): MMPS-005A, MMPS-005B