Learn More
Enzo Life Sciences MMP-10 (catalytic domain) (human), (recombinant) (10 µg)
SDP
Catalog No. 502011985
Encompass_Preferred
Alternative name: Matrix metalloproteinase 10, Stromelysin 2. Formulation: Liquid. In 50mM TRIS, 5mM CaCl2, 300mM NaCl, 20µM ZnCl2, 0.5% Brij-35, and 30% glycerol. Source: Produced in E. coli. Active Matrix Metalloproteinase-10 (MMP-10, stromelysin-2, transin-2) catalytic domain from human cDNA. The enzyme consists of the catalytic domain of human MMP-10 (Phe99-Glu271, NM_2425) with a C-terminal purification tag. This comprises an active form of MMP-10 which lacks the C-terminal hemopexin domain. MMPs lacking this domain cannot cleave native collagens; however, activity toward other targets such as gelatin, casein, or peptide substrates is unaffected. Long Term Storage: -80°C.
Catalog No. 50-201-1985 Supplier Enzo Life Sciences Supplier No. BMLSE3290010
Alternative name: Matrix metalloproteinase 10, Stromelysin 2. Formulation: Liquid. In 50mM TRIS, 5mM CaCl2, 300mM NaCl, 20µM ZnCl2, 0.5% Brij-35, and 30% glycerol. Source: Produced in E. coli. Active Matrix Metalloproteinase-10 (MMP-10, stromelysin-2, transin-2) catalytic domain from human cDNA. The enzyme consists of the catalytic domain of human MMP-10 (Phe99-Glu271, NM_2425) with a C-terminal purification tag. This comprises an active form of MMP-10 which lacks the C-terminal hemopexin domain. MMPs lacking this domain cannot cleave native collagens; however, activity toward other targets such as gelatin, casein, or peptide substrates is unaffected. Long Term Storage: -80°C.