Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MMP-10 protein, human recombinant (P. pastoris, His tag) | >90.0% | 100UG
SDP

Catalog No. 5000174435
Encompass_Preferred

Recombinant human MMP-10 supplied as a lyophilized, His-tagged protein expressed in Pichia pastoris for research applications. The product includes full sequence information, formulation and storage guidance; enzymatic activity is under evaluation and should not be assumed guaranteed.

  • Recombinant human MMP-10 expressed in Pichia pastoris.
  • N-terminal 6xHis tag for purification and detection.
  • Lyophilized powder formulated in 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose, pH 8.0.
  • Available in sizes up to 100 μg; store lyophilized at -20°C for long term.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Molecular weight approximately 45 kDa.
  • Enzymatic activity testing in progress; activity not guaranteed.
  • For research use only; reconstitute to ≥100 μg/mL in ddH2O as recommended.

Catalog No. 50-001-74435 Supplier Medchemexpress LLC Supplier No. HYP700571100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human MMP-10 supplied as a lyophilized, His-tagged protein expressed in Pichia pastoris for research applications. The product includes full sequence information, formulation and storage guidance; enzymatic activity is under evaluation and should not be assumed guaranteed.

  • Recombinant human MMP-10 expressed in Pichia pastoris.
  • N-terminal 6xHis tag for purification and detection.
  • Lyophilized powder formulated in 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose, pH 8.0.
  • Available in sizes up to 100 μg; store lyophilized at -20°C for long term.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Molecular weight approximately 45 kDa.
  • Enzymatic activity testing in progress; activity not guaranteed.
  • For research use only; reconstitute to ≥100 μg/mL in ddH2O as recommended.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.