Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Stromelysin-2 (matrix metalloproteinase 10) | >90.0% | 50UG
SDP

Catalog No. 5000175469
Encompass_Preferred

Recombinant human stromelysin-2 (MMP-10) expressed in Pichia pastoris with an N-terminal 6xHis tag. The lyophilized protein corresponds to residues F99-C476 (UniProt P09238), has a reported molecular weight of ~45 kDa, and is stated to be >90% pure by reducing SDS-PAGE. Supplied lyophilized in Tris-EDTA with trehalose; enzyme activity testing is in progress. Store frozen and aliquot to minimize freeze-thaw cycles.

  • Recombinant human protein expressed in Pichia pastoris.
  • Includes N-terminal 6xHis tag for purification.
  • Amino acid range corresponds to F99-C476 of UniProt P09238.
  • Molecular weight approximately 45 kDa and purity >90%.
  • Lyophilized formulation with trehalose for enhanced stability.
  • Store frozen and aliquot to avoid repeated freeze-thaw.

Catalog No. 50-001-75469 Supplier Medchemexpress LLC Supplier No. HYP70057150UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human stromelysin-2 (MMP-10) expressed in Pichia pastoris with an N-terminal 6xHis tag. The lyophilized protein corresponds to residues F99-C476 (UniProt P09238), has a reported molecular weight of ~45 kDa, and is stated to be >90% pure by reducing SDS-PAGE. Supplied lyophilized in Tris-EDTA with trehalose; enzyme activity testing is in progress. Store frozen and aliquot to minimize freeze-thaw cycles.

  • Recombinant human protein expressed in Pichia pastoris.
  • Includes N-terminal 6xHis tag for purification.
  • Amino acid range corresponds to F99-C476 of UniProt P09238.
  • Molecular weight approximately 45 kDa and purity >90%.
  • Lyophilized formulation with trehalose for enhanced stability.
  • Store frozen and aliquot to avoid repeated freeze-thaw.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.