Learn More
Enzo Life Sciences MMP-12 (CT) polyclonal antibody (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLSA4670100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to a portion of C-terminal mouse MMP-12 (matrix metalloproteinase 12). Species reactivity: Human, Mouse, Rat. Applications: WB. Formulation: Liquid. In 50% glycerol.
Catalog No. 50-201-1830