Learn More
Enzo Life Sciences MMP-12 (catalytic domain) (human), (recombinant) (10 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE1380010
Encompass_Preferred
Alternative name: Matrix metalloproteinase 12, Metalloelastase, Macrophage elastase. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 50mM TRIS, pH 9.5, containing 5mM calcium chloride, 500mM sodium chloride, 20µM zinc chloride, 0.5% Brij-35, and 30% glycerol. Source: Produced in E. coli. Active Matrix Metalloproteinase-12 (MMP-12, metalloelastase, macrophage elastase; often confused with neutrophil elastase) catalytic domain from human cDNA. The enzyme consists of the catalytic domain of human MMP-12 (Phe99-Leu271, NM_002426) with a C-terminal purification tag. Long Term Storage: -80°C.
Catalog No. 50-201-1940