Learn More
Enzo Life Sciences MMP-13 (human) polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ALX210833R100
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human MMP-13 (matrix metalloproteinase 13). UniProt ID: P45452. Species reactivity: Human. Creossreactivity: Does not cross-react with human MMP-2, MMP-9, MMP-14, MMP-15, MMP-16, MMP-17 or MMP-24. Applications: ELISA, WB. Recommended Dilutions: ELISA (detection, 1:2,000-1:4,000, use ALX-804-365 for capture) Western Blot (ECL-detection, 1:2,500 - 1:8,000) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS, pH 7.5, containing 0.01% thimerosal. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-8169