Learn More
Enzo Life Sciences MMP-13 (human), (recombinant) (10 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE4930010
Encompass_Preferred
Alternative name: Matrix metalloproteinase 13, Collagenase 3. Source: Produced in insect cells. Primarily full length latent MMP-13, with some active forms present. Produced in a baculovirus expression system. Long Term Storage: -80°C.
Catalog No. 50-201-2051